accessibility menu, dialog, popup

UserName

CAS RN 1192352-08-1

IUPAC Name: 4,7-dibromo-5,6-bis(octyloxy)-2,1,3-benzothiadiazole
Synonyms: 4,7-dibromo-5,6-bis(octyloxy)-2,1,3-benzothiadiazole
Molecular Weight (g/mol): 550.39
Molecular Formula: C22H34Br2N2O2S
InChi Key: VGCVJZVDAAWTFC-UHFFFAOYSA-N
SMILES: CCCCCCCCOC1=C(Br)C2=NSN=C2C(Br)=C1OCCCCCCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results