man

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 119-94-8

H3CNCH3

CAS RN 119-94-8

IUPAC Name: N-benzyl-N-ethyl-3-methylaniline
Synonyms: N-benzyl-N-ethyl-3-methylaniline
Molecular Weight (g/mol): 225.34
Molecular Formula: C16H19N
InChi Key: VIACAIAQHPLINF-UHFFFAOYSA-N
SMILES: CCN(CC1=CC=CC=C1)C1=CC=CC(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results