accessibility menu, dialog, popup

UserName

CAS RN 118757-11-2

IUPAC Name: 1-(benzenesulfonyl)-5-bromo-1H-indole
Synonyms: 1-(benzenesulfonyl)-5-bromoindole
Molecular Weight (g/mol): 336.2
Molecular Formula: C14H10BrNO2S
InChi Key: XUKSXJWRFMTAPZ-UHFFFAOYSA-N
SMILES: BrC1=CC=C2N(C=CC2=C1)S(=O)(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results