accessibility menu, dialog, popup

UserName

H3CNH2(R)(R)H2NCH3

CAS RN 118628-68-5

IUPAC Name: methyl[(1R,2R)-2-(methylazaniumyl)-1,2-diphenylethyl]azanium
Synonyms: methyl[(1R,2R)-2-(methylammonio)-1,2-diphenylethyl]azanium
Molecular Weight (g/mol): 242.37
Molecular Formula: C16H22N2
InChi Key: BTHYVQUNQHWNLS-HZPDHXFCSA-P
SMILES: C[NH2+][C@@H]([C@H]([NH2+]C)C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results