accessibility menu, dialog, popup

UserName

CAS RN 118525-40-9

CH3OHOOOHOHCH3H3CO

CAS RN 118525-40-9

IUPAC Name: 3,5,7-trihydroxy-2-(4-methoxyphenyl)-8-(3-methylbut-2-en-1-yl)-4H-chromen-4-one
Synonyms: 3,5,7-trihydroxy-2-(4-methoxyphenyl)-8-(3-methylbut-2-en-1-yl)chromen-4-one
Molecular Weight (g/mol): 368.39
Molecular Formula: C21H20O6
InChi Key: TUUXBSASAQJECY-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1=C(O)C(=O)C2=C(O)C=C(O)C(CC=C(C)C)=C2O1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results