accessibility menu, dialog, popup

UserName

Explore All Our Special Offers, All in One Place

Save Today

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 117903-53-4

H3N(R)(S)NH3BrBr

CAS RN 117903-53-4

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: —
InChi Key: —
SMILES: —

Narrow Results

Narrow Results

  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.