accessibility menu, dialog, popup

Test

CAS RN 117724-63-7

CAS RN 117724-63-7

IUPAC Name: 2-methyl-4-(trifluoromethyl)-1,3-thiazole-5-carboxylic acid
Synonyms: 2-methyl-4-(trifluoromethyl)-1,3-thiazole-5-carboxylic acid
Molecular Weight (g/mol): 211.16
Molecular Formula: C6H4F3NO2S
InChi Key: REKJPVUFKQYMHW-UHFFFAOYSA-N
SMILES: CC1=NC(=C(S1)C(O)=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results