CAS RN 117591-20-5
IUPAC Name:
benzyl N-[(1S)-3-methyl-1-{[(2S)-1-oxohexan-2-yl]carbamoyl}butyl]carbamate
Synonyms:
benzyl N-[(1S)-3-methyl-1-{[(2S)-1-oxohexan-2-yl]carbamoyl}butyl]carbamate
Molecular Weight (g/mol):
362.47
Molecular Formula:
C20H30N2O4
InChi Key:
PGGUOGKHUUUWAF-ROUUACIJSA-N
SMILES:
CCCC[C@H](NC(=O)[C@H](CC(C)C)NC(=O)OCC1=CC=CC=C1)C=O
1
–
1
of
1
results
1
Thermo Scientific Chemicals Calpeptin, ≥98%
CAS: 117591-20-5 Molecular Formula: C20H30N2O4 Synonym: N-Benzyloxycarbonyl-L-leucyl-norleucinal; Z-Leu-norleucinal