CAS RN 1174157-65-3
IUPAC Name:
2,5-dioxopyrrolidin-1-yl 3-(prop-2-yn-1-yloxy)propanoate
Synonyms:
2,5-dioxopyrrolidin-1-yl 3-(prop-2-yn-1-yloxy)propanoate
Molecular Weight (g/mol):
225.2
Molecular Formula:
C10H11NO5
InChi Key:
WKIKHHMUNOVQLD-UHFFFAOYSA-N
SMILES:
O=C(CCOCC#C)ON1C(=O)CCC1=O
1
–
1
of
1
results
1
N-Succinimidyl 3-(propargyloxy)propionate
CAS: 1174157-65-3 Molecular Formula: C10H11NO5 Molecular Weight (g/mol): 225.20 MDL Number: MFCD13184973 InChI Key: WKIKHHMUNOVQLD-UHFFFAOYSA-N Synonym: n-succinimidyl 3-propargyloxy propionate,propargyl-nhs,ambotzpeg1935,acetylene-peg-nhs ester,propargyl-peg1-nhs ester,propargyl-succinimidyl-ester,propargyl-n-hydroxysuccinimidyl ester,2,5-dioxopyrrolidin-1-yl 3-prop-2-ynyloxy propanoate,2,5-dioxopyrrolidin-1-yl 3-prop-2-yn-1-yloxy propanoate PubChem CID: 51340986 IUPAC Name: 2,5-dioxopyrrolidin-1-yl 3-(prop-2-yn-1-yloxy)propanoate SMILES: O=C(CCOCC#C)ON1C(=O)CCC1=O