accessibility menu, dialog, popup

UserName

CAS RN 117098-93-8

IUPAC Name: 6-amino-1,3-dihydro-2,1-benzoxaborol-1-ol hydrochloride
Synonyms: 6-amino-3H-2,1-benzoxaborol-1-ol hydrochloride
Molecular Weight (g/mol): 185.41
Molecular Formula: C7H9BClNO2
InChi Key: ZDCBDYGPSUVCOU-UHFFFAOYSA-N
SMILES: Cl.NC1=CC2=C(COB2O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results