accessibility menu, dialog, popup

UserName

OOHH2NHSH2OHCl

CAS RN 116797-51-4

IUPAC Name: 2-amino-3-sulfanylpropanoic acid hydrate hydrochloride
Synonyms: cysteine hydrate hydrochloride
Molecular Weight (g/mol): 175.63
Molecular Formula: C3H10ClNO3S
InChi Key: QIJRTFXNRTXDIP-UHFFFAOYNA-N
SMILES: O.Cl.NC(CS)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results