accessibility menu, dialog, popup

UserName

H3CCH3OSSOCH3CH3

CAS RN 1160823-77-7

IUPAC Name: 2,8-bis[(2-ethylhexyl)oxy]-4,10-dithiatricyclo[7.3.0.0³,⁷]dodeca-1,3(7),5,8,11-pentaene
Synonyms: 2,8-bis[(2-ethylhexyl)oxy]-4,10-dithiatricyclo[7.3.0.0³,⁷]dodeca-1,3(7),5,8,11-pentaene
Molecular Weight (g/mol): 446.71
Molecular Formula: C26H38O2S2
InChi Key: HATOWNJGYIVNBU-UHFFFAOYNA-N
SMILES: CCCCC(CC)COC1=C2C=CSC2=C(OCC(CC)CCCC)C2=C1SC=C2
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results