CAS RN 116-95-0
CAS RN 116-95-0
IUPAC Name:
bis[4-(diethylamino)phenyl](2-sulfo-4-sulfonatophenyl)methylium
Synonyms:
bis[4-(diethylamino)phenyl](2-sulfo-4-sulfonatophenyl)methylium
Molecular Weight (g/mol):
544.68
Molecular Formula:
C27H32N2O6S2
InChi Key:
IKHKJYWPWWBSFZ-UHFFFAOYSA-N
SMILES:
CCN(CC)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC)C1=CC=C(C=C1S(O)(=O)=O)S([O-])(=O)=O
1
–
1
of
1
results
1
Acid Blue 1, TCI Americaâ„¢
CAS: 116-95-0 Molecular Formula: C27H32N2O6S2 Molecular Weight (g/mol): 544.681 MDL Number: MFCD00012119 InChI Key: IKHKJYWPWWBSFZ-UHFFFAOYSA-N Synonym: Alphazurine G, Brilliant Acid Blue, Carmine Blue V, Patent Blue V, Xylene Blue, Azure Blue VX PubChem CID: 4294647 IUPAC Name: 2-[[4-(diethylamino)phenyl]-(4-diethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]-5-sulfobenzenesulfonate SMILES: CCN(CC)C1=CC=C(C=C1)C(=C2C=CC(=[N+](CC)CC)C=C2)C3=C(C=C(C=C3)S(=O)(=O)O)S(=O)(=O)[O-]