accessibility menu, dialog, popup

UserName

NHOPOOHHOPOHOONa

CAS RN 115436-72-1

IUPAC Name: sodium hydrogen [1-hydroxy-1-phosphono-2-(pyridin-3-yl)ethyl]phosphonate
Synonyms: sodium hydrogen 1-hydroxy-1-phosphono-2-(pyridin-3-yl)ethylphosphonate
Molecular Weight (g/mol): 305.09
Molecular Formula: C7H10NNaO7P2
InChi Key: DRFDPXKCEWYIAW-UHFFFAOYNA-M
SMILES: [Na+].OC(CC1=CN=CC=C1)(P(O)(O)=O)P(O)([O-])=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)