accessibility menu, dialog, popup

UserName

CAS RN 113808-86-9

IUPAC Name: 3,5-dimethyl-1H-pyrazole-4-carboxylic acid
Synonyms: 3,5-dimethyl-1H-pyrazole-4-carboxylic acid
Molecular Weight (g/mol): 140.14
Molecular Formula: C6H8N2O2
InChi Key: IOOWDXMXZBYKLR-UHFFFAOYSA-N
SMILES: CC1=C(C(O)=O)C(C)=NN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results