accessibility menu, dialog, popup

UserName

SSSSSSSS

CAS RN 113728-71-5

IUPAC Name: 5-{[2,2'-bithiophen]-5-yl}-5'-(5'-{[2,2'-bithiophen]-5-yl}-[2,2'-bithiophen]-5-yl)-2,2'-bithiophene
Synonyms: 5-{[2,2'-bithiophen]-5-yl}-5'-(5'-{[2,2'-bithiophen]-5-yl}-[2,2'-bithiophen]-5-yl)-2,2'-bithiophene
Molecular Weight (g/mol): 658.98
Molecular Formula: C32H18S8
InChi Key: GCMCTPRNKVKGCY-UHFFFAOYSA-N
SMILES: S1C=CC=C1C1=CC=C(S1)C1=CC=C(S1)C1=CC=C(S1)C1=CC=C(S1)C1=CC=C(S1)C1=CC=C(S1)C1=CC=CS1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results