accessibility menu, dialog, popup

UserName

ClH3COO(R)(S)H2NH

CAS RN 1127-99-7

IUPAC Name: hydrogen ethyl (1R,2S)-2-aminocyclohexane-1-carboxylate chloride
Synonyms: hydrogen ethyl (1R,2S)-2-aminocyclohexane-1-carboxylate chloride
Molecular Weight (g/mol): 207.7
Molecular Formula: C9H18ClNO2
InChi Key: XMQSOBPCWYVZSW-WLYNEOFISA-N
SMILES: [H+].[Cl-].CCOC(=O)[C@@H]1CCCC[C@@H]1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)