CAS RN 1126-74-5
IUPAC Name:
(2E)-3-(pyridin-3-yl)prop-2-enoate
Synonyms:
(2E)-3-(pyridin-3-yl)prop-2-enoate
Molecular Weight (g/mol):
148.14
Molecular Formula:
C8H6NO2
InChi Key:
VUVORVXMOLQFMO-ONEGZZNKSA-M
SMILES:
[O-]C(=O)\C=C\C1=CC=CN=C1
1
–
1
of
1
results
1
3-(3-Pyridyl)acrylic Acid 98.0+%, TCI America™
CAS: 1126-74-5 Molecular Formula: C8H7NO2 Molecular Weight (g/mol): 149.149 MDL Number: MFCD00006410 InChI Key: VUVORVXMOLQFMO-ONEGZZNKSA-N Synonym: 3-3-pyridyl acrylic acid,trans-3-3-pyridyl acrylic acid,e-3-pyridin-3-yl acrylic acid,3-pyridylacrylic acid,2e-3-pyridin-3-ylacrylic acid,2e-3-pyridin-3-yl prop-2-enoic acid,3-pyridin-3-yl acrylic acid,trans-3-3'-pyridyl acrylic acid,3-3-pyridinyl acrylic acid,3-pyridinyl-acrylic acid PubChem CID: 776396 IUPAC Name: (E)-3-pyridin-3-ylprop-2-enoic acid SMILES: C1=CC(=CN=C1)C=CC(=O)O