CAS RN 112245-09-7
IUPAC Name:
(2R)-2-amino-3,3-dimethylbutan-1-ol
Synonyms:
(2R)-2-amino-3,3-dimethylbutan-1-ol
Molecular Weight (g/mol):
117.19
Molecular Formula:
C6H15NO
InChi Key:
JBULSURVMXPBNA-MLHKIVSYNA-N
SMILES:
CC(C)(C)[C@@H](N)CO
1
–
1
of
1
results
1
Promotions Available
(R)-(-)-tert-Leucinol, 98%, Thermo Scientific™
(R)-(-)-tert-Leucinol, 98%, Thermo Scientific™
CAS: 112245-09-7 Molecular Formula: C6H15NO Molecular Weight (g/mol): 117.192 InChI Key: JBULSURVMXPBNA-YFKPBYRVSA-N Synonym: r-tert-leucinol,d-tert-leucinol,2r-2-amino-3,3-dimethylbutan-1-ol,r-2-amino-3,3-dimethylbutan-1-ol,r-2-amino-3,3-dimethyl-1-butanol,2r-2-amino-3,3-dimethyl-1-butanol,r---tert-leucinol,?-tert-leucinol,r-?-tert-leucinol PubChem CID: 6932847 IUPAC Name: (2R)-2-amino-3,3-dimethylbutan-1-ol SMILES: CC(C)(C)C(CO)N