CAS RN 1120-88-3
IUPAC Name:
4-methylpyridazine
Synonyms:
pyridazine, 4-methyl-
Molecular Weight (g/mol):
94.12
Molecular Formula:
C5H6N2
InChi Key:
AIKUBOPKWKZULG-UHFFFAOYSA-N
SMILES:
CC1=CC=NN=C1
1
–
1
of
1
results
1
4-Methylpyridazine 98.0+%, TCI America™
CAS: 1120-88-3 Molecular Formula: C5H6N2 Molecular Weight (g/mol): 94.117 MDL Number: MFCD00011594 InChI Key: AIKUBOPKWKZULG-UHFFFAOYSA-N Synonym: pyridazine, 4-methyl,4-methyl pyridazine,4-methyl-1,2-diazine,4-methylpyridazine,acmc-209z0j,ksc183s2h,1,2-oxathiolane 2,2-dioxide; 3-hydroxy-1-propanesulfonic acid gamma-sultone; propane sultone PubChem CID: 136882 IUPAC Name: 4-methylpyridazine SMILES: CC1=CN=NC=C1