accessibility menu, dialog, popup

UserName

CH3PH3CH3CH3CFPFFFFF

CAS RN 111928-07-5

IUPAC Name: hexafluoro-λ⁵-phosphanuide; tetraethylphosphanium
Synonyms: tetraethylphosphanium hexafluorophosphate
Molecular Weight (g/mol): 292.19
Molecular Formula: C8H20F6P2
InChi Key: WJZBUQKNTCKXAE-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.CC[P+](CC)(CC)CC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results