accessibility menu, dialog, popup

UserName

Uncover New Spotlight Deals and Get Up to 50% Off

Save Today

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 1117-19-7

H3COOOOCH3

CAS RN 1117-19-7

IUPAC Name: 1,3-dipropyl propanedioate
Synonyms: 1,3-dipropyl propanedioate
Molecular Weight (g/mol): 188.22
Molecular Formula: C9H16O4
InChi Key: LWIWFCDNJNZEKB-UHFFFAOYSA-N
SMILES: CCCOC(=O)CC(=O)OCCC
Boiling Point 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.