accessibility menu, dialog, popup

UserName

NHNNH2

CAS RN 111562-32-4

IUPAC Name: 2-(1H-pyrazol-5-yl)aniline
Synonyms: 2-(2H-pyrazol-3-yl)aniline
Molecular Weight (g/mol): 159.19
Molecular Formula: C9H9N3
InChi Key: NRSKRRKWPPDKNI-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1C1=CC=NN1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results