accessibility menu, dialog, popup

UserName

FClN

CAS RN 110888-15-8

IUPAC Name: 4-chloro-3-fluorobenzonitrile
Synonyms: 4-chloro-3-fluorobenzonitrile
Molecular Weight (g/mol): 155.56
Molecular Formula: C7H3ClFN
InChi Key: GWZQVECNESCSKR-UHFFFAOYSA-N
SMILES: FC1=CC(=CC=C1Cl)C#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results