CAS RN 110871-86-8
IUPAC Name:
5-amino-1-cyclopropyl-7-[(3R,5S)-3,5-dimethylpiperazin-1-yl]-6,8-difluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
Synonyms:
sparfloxacin zagam sparfloxacinum sparfloxacine esparfloxacino SPFX
Molecular Weight (g/mol):
392.41
Molecular Formula:
C19H22F2N4O3
InChi Key:
DZZWHBIBMUVIIW-DTORHVGOSA-N
SMILES:
C[C@H]1CN(C[C@@H](C)N1)C1=C(F)C(N)=C2C(=O)C(=CN(C3CC3)C2=C1F)C(O)=O
1
–
1
of
1
results
1
Sparfloxacin, 98%
CAS: 110871-86-8 Molecular Formula: C19H22F2N4O3 Molecular Weight (g/mol): 392.41 MDL Number: MFCD00869619 InChI Key: DZZWHBIBMUVIIW-DTORHVGOSA-N Synonym: sparfloxacin,zagam,esparfloxacino,sparfloxacine,sparfloxacinum,spara,sparfloxacine inn-french,sparfloxacinum inn-latin,spfx,esparfloxacino inn-spanish PubChem CID: 60464 ChEBI: CHEBI:9212 IUPAC Name: 5-amino-1-cyclopropyl-7-[(3R,5S)-3,5-dimethylpiperazin-1-yl]-6,8-difluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid SMILES: C[C@H]1CN(C[C@@H](C)N1)C1=C(F)C(N)=C2C(=O)C(=CN(C3CC3)C2=C1F)C(O)=O