accessibility menu, dialog, popup

UserName

H3CCH3OO

CAS RN 110-45-2

IUPAC Name: 3-methylbutyl formate
Synonyms: isoamyl formate
Molecular Weight (g/mol): 116.16
Molecular Formula: C6H12O2
InChi Key: XKYICAQFSCFURC-UHFFFAOYSA-N
SMILES: CC(C)CCOC=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results