accessibility menu, dialog, popup

UserName

H3CNNNNFNONH

CAS RN 108612-45-9

IUPAC Name: 2-[(1-{1-[(4-fluorophenyl)methyl]-1H-1,3-benzodiazol-2-yl}piperidin-4-yl)(methyl)amino]-3,4-dihydropyrimidin-4-one
Synonyms: mizolastine
Molecular Weight (g/mol): 432.5
Molecular Formula: C24H25FN6O
InChi Key: PVLJETXTTWAYEW-UHFFFAOYSA-N
SMILES: CN(C1CCN(CC1)C1=NC2=CC=CC=C2N1CC1=CC=C(F)C=C1)C1=NC=CC(=O)N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results