accessibility menu, dialog, popup

UserName

CH3OOOCH3CH3CH3CH3CH3

CAS RN 108031-79-4

IUPAC Name: 1,10,10-trimethyl-4-({1,10,10-trimethyl-3-oxatricyclo[5.2.1.0²,⁶]decan-4-yl}oxy)-3-oxatricyclo[5.2.1.0²,⁶]decane
Synonyms: 1,10,10-trimethyl-4-({1,10,10-trimethyl-3-oxatricyclo[5.2.1.0²,⁶]decan-4-yl}oxy)-3-oxatricyclo[5.2.1.0²,⁶]decane
Molecular Weight (g/mol): 374.57
Molecular Formula: C24H38O3
InChi Key: VUDXCBLBKXFCNA-UHFFFAOYNA-N
SMILES: CC1(C)C2CCC1(C)C1OC(CC21)OC1CC2C3CCC(C)(C2O1)C3(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results