accessibility menu, dialog, popup

UserName

CAS RN 1076-28-4

IUPAC Name: 2-methylquinolin-1-ium-1-olate
Synonyms: 2-methylquinolin-1-ium-1-olate
Molecular Weight (g/mol): 159.19
Molecular Formula: C10H9NO
InChi Key: FZJUONUBFWNHNU-UHFFFAOYSA-N
SMILES: CC1=CC=C2C=CC=CC2=[N+]1[O-]
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results