accessibility menu, dialog, popup

UserName

CH3ONH2N

CAS RN 1072-67-9

IUPAC Name: 5-methyl-1,2-oxazol-3-amine
Synonyms: 5-methyl-1,2-oxazol-3-amine
Molecular Weight (g/mol): 98.11
Molecular Formula: C4H6N2O
InChi Key: FKPXGNGUVSHWQQ-UHFFFAOYSA-N
SMILES: CC1=CC(N)=NO1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results