accessibility menu, dialog, popup

UserName

CAS RN 1072-52-2

OHN

CAS RN 1072-52-2

IUPAC Name: 2-(aziridin-1-yl)ethan-1-ol
Synonyms: 1-aziridineethanol
Molecular Weight (g/mol): 87.12
Molecular Formula: C4H9NO
InChi Key: VYONOYYDEFODAJ-UHFFFAOYSA-N
SMILES: OCCN1CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results