accessibility menu, dialog, popup

UserName

Check Out the New Spotlight Offers and Get Up to 50% Off

Save Today

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 1069-23-4

OHH2COHCH2

CAS RN 1069-23-4

IUPAC Name: hexa-1,5-diene-3,4-diol
Synonyms: 1,5-hexadiene,3,4-dihydroxy-
Molecular Weight (g/mol): 114.14
Molecular Formula: C6H10O2
InChi Key: KUQWZSZYIQGTHT-UHFFFAOYNA-N
SMILES: OC(C=C)C(O)C=C
Boiling Point 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.