CAS RN 106876-54-4
IUPAC Name:
2,2-difluoro-2H-1,3-benzodioxol-4-amine
Synonyms:
2,2-difluoro-1,3-benzodioxol-4-amine
Molecular Weight (g/mol):
173.12
Molecular Formula:
C7H5F2NO2
InChi Key:
RXIKYXZKSIARLN-UHFFFAOYSA-N
SMILES:
NC1=CC=CC2=C1OC(F)(F)O2
1
–
1
of
1
results
1
Promotions Available
4-Amino-2,2-difluoro-1,3-benzodioxole, 97+%
4-Amino-2,2-difluoro-1,3-benzodioxole, 97+%
CAS: 106876-54-4 Molecular Formula: C7H5F2NO2 Molecular Weight (g/mol): 173.12 MDL Number: MFCD00792416 InChI Key: RXIKYXZKSIARLN-UHFFFAOYSA-N Synonym: 4-amino-2,2-difluoro-1,3-benzodioxole,2,2-difluorobenzo d 1,3 dioxol-4-amine,2,3-difluoromethylenedioxy aniline,2,2-difluoro-4-amino-1,3-benzodioxole,1,3-benzodioxol-4-amine, 2,2-difluoro,2,2-difluoro-2h-1,3-benzodioxol-4-amine,2,2-difluoro-benzo 1,3 dioxol-4-ylamine,2,3-difluoromethylenedioxy-aniline,acmc-1btze,2,2-difluoro-1,3-benzodioxole-4-amine PubChem CID: 2736891 IUPAC Name: 2,2-difluoro-2H-1,3-benzodioxol-4-amine SMILES: NC1=CC=CC2=C1OC(F)(F)O2