accessibility menu, dialog, popup

UserName

BrHSnHCH3CH3H3C

CAS RN 1066-44-0

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 245.74
Molecular Formula: C3H11BrSn
InChi Key: KNPKGRQZYPSMBF-UHFFFAOYSA-N
SMILES: Br.C[SnH](C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results