accessibility menu, dialog, popup

UserName

H3COOOOCH3

CAS RN 106-19-4

IUPAC Name: 1,6-dipropyl hexanedioate
Synonyms: dipropyl adipate
Molecular Weight (g/mol): 230.3
Molecular Formula: C12H22O4
InChi Key: NKOUWLLFHNBUDW-UHFFFAOYSA-N
SMILES: CCCOC(=O)CCCCC(=O)OCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results