accessibility menu, dialog, popup

UserName

OO(S)H2N

CAS RN 105400-81-5

IUPAC Name: (1S)-1-(carboxylatomethyl)-1,2,3,4-tetrahydroisoquinolin-2-ium
Synonyms: (1S)-1-(carboxylatomethyl)-1,2,3,4-tetrahydroisoquinolin-2-ium
Molecular Weight (g/mol): 191.23
Molecular Formula: C11H13NO2
InChi Key: BUAVPRGEIAVFBF-JTQLQIEISA-N
SMILES: [O-]C(=O)C[C@@H]1[NH2+]CCC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)