CAS RN 105264-63-9
CAS RN 105264-63-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-(8-Hydroxyoctyl)phthalimide 98.0+%, TCI America™
CAS: 105264-63-9 Molecular Formula: C16H21NO3 Molecular Weight (g/mol): 275.348 MDL Number: MFCD00191517 InChI Key: RBDXDFOXVNNMNG-UHFFFAOYSA-N Synonym: 8-(N-Phthalimidyl)-1-octanol PubChem CID: 21884230 IUPAC Name: 2-(8-hydroxyoctyl)isoindole-1,3-dione SMILES: C1=CC=C2C(=C1)C(=O)N(C2=O)CCCCCCCCO