accessibility menu, dialog, popup

UserName

CH3SiH3CCH2H2C

CAS RN 10519-87-6

IUPAC Name: diethenyldimethylsilane
Synonyms: diethenyldimethylsilane
Molecular Weight (g/mol): 112.25
Molecular Formula: C6H12Si
InChi Key: QRHCILLLMDEFSD-UHFFFAOYSA-N
SMILES: C[Si](C)(C=C)C=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results