CAS RN 10514-70-2
IUPAC Name:
2,2-dimethyl-3,4-dihydro-2H-1,4-benzoxazin-3-one
Synonyms:
2,2-dimethyl-4H-1,4-benzoxazin-3-one
Molecular Weight (g/mol):
177.2
Molecular Formula:
C10H11NO2
InChi Key:
LJRUKUQEIHJTEL-UHFFFAOYSA-N
SMILES:
CC1(C)OC2=CC=CC=C2NC1=O
1
–
1
of
1
results
1
Promotions Available
2,2-Dimethyl-2H-1,4-benzoxazin-3(4H)-one, 97%, Thermo Scientific™
2,2-Dimethyl-2H-1,4-benzoxazin-3(4H)-one, 97%, Thermo Scientific™
CAS: 10514-70-2 Molecular Formula: C10H11NO2 Molecular Weight (g/mol): 177.20 MDL Number: MFCD11868710 InChI Key: LJRUKUQEIHJTEL-UHFFFAOYSA-N Synonym: 2,2-dimethyl-2h-benzo b 1,4 oxazin-3 4h-one,2h-1,4-benzoxazin-3 4h-one, 2,2-dimethyl,2,2-dimethyl-3,4-dihydro-2h-1,4-benzoxazin-3-one,2,2-dimethyl-1,4-benzoxazin-3-one,2,2-dimethyl-4h-benzo 1,4 oxazin-3-one,2,2-dimethyl-2h-1,4-benzoxazin-3 4h-one,2,2-dimethyl-2,3-dihydro-4h-1,4-benzooxazine-3-one PubChem CID: 10607395 IUPAC Name: 2,2-dimethyl-4H-1,4-benzoxazin-3-one SMILES: CC1(C)OC2=CC=CC=C2NC1=O