accessibility menu, dialog, popup

UserName

CAS RN 10500-34-2

H3COOO(E)

CAS RN 10500-34-2

IUPAC Name: 3-[(2E)-hex-2-en-1-yl]oxolane-2,5-dione
Synonyms: 3-[(2E)-hex-2-en-1-yl]oxolane-2,5-dione
Molecular Weight (g/mol): 182.22
Molecular Formula: C10H14O3
InChi Key: VWXVGDSPDMSWFP-SNAWJCMRNA-N
SMILES: CCC\C=C\CC1CC(=O)OC1=O
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results