accessibility menu, dialog, popup

UserName

OPSOONaNaNa

CAS RN 10489-48-2

IUPAC Name: trisodium phosphorothioate
Synonyms: trisodium thiophosphate
Molecular Weight (g/mol): 180
Molecular Formula: Na3O3PS
InChi Key: RIFYBBXGYKFBFC-UHFFFAOYSA-K
SMILES: [Na+].[Na+].[Na+].[O-]P([O-])([O-])=S
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)