accessibility menu, dialog, popup

Test

CAS RN 104863-67-4

CAS RN 104863-67-4

IUPAC Name: 2,2,2-trifluoro-N-methoxy-N-methylacetamide
Synonyms: 2,2,2-trifluoro-N-methoxy-N-methylacetamide
Molecular Weight (g/mol): 157.09
Molecular Formula: C4H6F3NO2
InChi Key: JDBJZZTZYKUFFK-UHFFFAOYSA-N
SMILES: CON(C)C(=O)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results