accessibility menu, dialog, popup

UserName

CH3NNSH3C

CAS RN 104208-13-1

IUPAC Name: 1,3-dimethyl-2-(thiophen-2-yl)imidazolidine
Synonyms: 1,3-dimethyl-2-(thiophen-2-yl)imidazolidine
Molecular Weight (g/mol): 182.29
Molecular Formula: C9H14N2S
InChi Key: HIHONCXGFIYIJX-UHFFFAOYSA-N
SMILES: CN1CCN(C)C1C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results