accessibility menu, dialog, popup

UserName

CAS RN 1041005-52-0

IUPAC Name: dichloropalladium; dicyclohexyl[3-(dicyclohexylphosphaniumyl)propyl]phosphanium
Synonyms: dicyclohexyl[3-(dicyclohexylphosphaniumyl)propyl]phosphanium; palladium chloride
Molecular Weight (g/mol): 615.98
Molecular Formula: C27H52Cl2P2Pd
InChi Key: JLEVAMQVDDHWOI-UHFFFAOYSA-N
SMILES: Cl[Pd]Cl.C(C[PH+](C1CCCCC1)C1CCCCC1)C[PH+](C1CCCCC1)C1CCCCC1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)