accessibility menu, dialog, popup

UserName

CAS RN 10377-52-3

LiLiLiOPOOO

CAS RN 10377-52-3

IUPAC Name: trilithium(1+) phosphate
Synonyms: lithium phosphate
Molecular Weight (g/mol): 115.79
Molecular Formula: Li3O4P
InChi Key: TWQULNDIKKJZPH-UHFFFAOYSA-K
SMILES: [Li+].[Li+].[Li+].[O-]P([O-])([O-])=O
Grade 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)