accessibility menu, dialog, popup

UserName

H3CCH3OOCH3(E)

CAS RN 10371-45-6

IUPAC Name: butan-2-yl (2E)-but-2-enoate
Synonyms: sec-butyl (2E)-but-2-enoate
Molecular Weight (g/mol): 142.2
Molecular Formula: C8H14O2
InChi Key: KAKJZWSUVDRWQU-GQCTYLIANA-N
SMILES: CCC(C)OC(=O)\C=C\C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results