accessibility menu, dialog, popup

UserName

OOOHH2N

CAS RN 10366-71-9

IUPAC Name: 2-(2-aminoethoxy)acetic acid
Synonyms: (2-aminoethoxy)acetic acid
Molecular Weight (g/mol): 119.12
Molecular Formula: C4H9NO3
InChi Key: GNRLUBOJIGSVNT-UHFFFAOYSA-N
SMILES: NCCOCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)