accessibility menu, dialog, popup

UserName

CAS RN 1034566-13-6

IUPAC Name: 2-methyl-4-(trifluoromethyl)-1,3-thiazole-5-carbaldehyde
Synonyms: 2-methyl-4-(trifluoromethyl)-1,3-thiazole-5-carbaldehyde
Molecular Weight (g/mol): 195.16
Molecular Formula: C6H4F3NOS
InChi Key: FBQFBBGXQZDOQV-UHFFFAOYSA-N
SMILES: CC1=NC(=C(S1)C=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)