accessibility menu, dialog, popup

UserName

CH3OOF

CAS RN 103438-88-6

IUPAC Name: 2-fluoro-3-methoxybenzaldehyde
Synonyms: 2-fluoro-3-methoxybenzaldehyde
Molecular Weight (g/mol): 154.14
Molecular Formula: C8H7FO2
InChi Key: LIHCOUDNHILORI-UHFFFAOYSA-N
SMILES: COC1=CC=CC(C=O)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)