accessibility menu, dialog, popup

Test

CAS RN 10220-46-9

CAS RN 10220-46-9

IUPAC Name: octadecyl 2-sulfanylacetate
Synonyms: octadecyl 2-sulfanylacetate
Molecular Weight (g/mol): 344.6
Molecular Formula: C20H40O2S
InChi Key: JQTFPHLEQLLQOT-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCOC(=O)CS
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results